ENA311760982 Enamine

2-(5-ethoxy-2-methyl-2,3-dihydro-1-benzofuran-6-yl)cyclopropane-1-carboxylic acid

  • MDL number MFCD12719526
  • Cas Number : 2059923-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102...104
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71881

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71881
Availability details for
Quantity
  • Availability
  • Add to Cart