ENA311760984 Enamine

2-fluoro-4,5-dimethoxyaniline

  • MDL number MFCD12174785
  • Cas Number : 397-75-1
  • Molecular Weight : 171.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-71883

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71883
Availability details for
Quantity
  • Availability
  • Add to Cart