ENA311760985 Enamine

2-(3,4-difluorophenoxy)-2-phenylacetic acid

  • MDL number MFCD11580097
  • Cas Number : 1082558-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71884

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71884
Availability details for
Quantity
  • Availability
  • Add to Cart