ENA311760998 Enamine

1-[4-(prop-2-yn-1-yloxy)phenyl]ethan-1-ol

  • MDL number MFCD12185868
  • Cas Number : 1156181-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71888

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71888
Availability details for
Quantity
  • Availability
  • Add to Cart