ENA311761000 Enamine

4-(2-cyano-3-fluorophenoxy)benzoic acid

  • MDL number MFCD07432805
  • Cas Number : 944663-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186...188
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71598

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71598
Availability details for
Quantity
  • Availability
  • Add to Cart