ENA311761006 Enamine

3-[5-(trifluoromethyl)-1,3-benzothiazol-2-yl]propanoic acid

  • MDL number MFCD08271870
  • Cas Number : 885527-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-71901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71901
Availability details for
Quantity
  • Availability
  • Add to Cart