ENA311761028 Enamine

1-(2,3-dihydro-1,4-benzodioxin-6-yl)-2-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD09947168
  • Cas Number : 1017417-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71915

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71915
Availability details for
Quantity
  • Availability
  • Add to Cart