ENA311761037 Enamine

2-(3,5-dimethyl-1,2-oxazol-4-yl)ethan-1-amine

  • MDL number MFCD03043478
  • Cas Number : 510717-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71921

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-71921
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart