ENA311761042 Enamine

3-(3-formylphenoxymethyl)benzoic acid

  • MDL number MFCD12544993
  • Cas Number : 479578-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122...124
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71932

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71932
Availability details for
Quantity
  • Availability
  • Add to Cart