ENA311761044 Enamine

5-(4-methoxyphenyl)-3-oxopentanenitrile

  • MDL number MFCD11187382
  • Cas Number : 1000895-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58...60
Purity 95.00
Storage temp. -20
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71934

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71934
Availability details for
Quantity
  • Availability
  • Add to Cart