ENA311761045 Enamine

5-(5-bromo-2-fluorophenyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD11137502
  • Cas Number : 299441-49-9
  • Molecular Weight : 274.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-71935

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71935
Availability details for
Quantity
  • Availability
  • Add to Cart