ENA311761046 Enamine

2-cyclopropyl-6-methylpyrimidin-4-ol

  • MDL number MFCD11054233
  • Molecular Weight : 150.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-71936

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71936
Availability details for
Quantity
  • Availability
  • Add to Cart