ENA311761064 Enamine

3-[1-(3,4-dichlorophenyl)-2,5-dioxoimidazolidin-4-yl]propanoic acid

  • MDL number MFCD13744125
  • Cas Number : 1192227-67-0
  • Molecular Weight : 317.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-71945

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71945
Availability details for
Quantity
  • Availability
  • Add to Cart