ENA311761067 Enamine

N-(2-aminophenyl)-2-(cyclopent-2-en-1-yl)acetamide

  • MDL number MFCD12084053
  • Cas Number : 1153541-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71946

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71946
Availability details for
Quantity
  • Availability
  • Add to Cart