ENA311761069 Enamine

2-(4-chloro-2-hydroxybenzamido)thiophene-3-carboxylic acid

  • MDL number MFCD12443778
  • Cas Number : 1041557-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-71601

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71601
Availability details for
Quantity
  • Availability
  • Add to Cart