ENA311761080 Enamine

1-(2-methanesulfonylphenyl)-1H-pyrazol-4-amine

  • MDL number MFCD14594690
  • Cas Number : 1250582-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71550

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71550
Availability details for
Quantity
  • Availability
  • Add to Cart