ENA311761083 Enamine

2-(2,4-dichlorophenyl)cyclopropane-1-carboxylic acid

  • MDL number MFCD12068264
  • Cas Number : 1157554-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71958

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71958
Availability details for
Quantity
  • Availability
  • Add to Cart