ENA311761088 Enamine

1-[(3-bromophenyl)carbonyl]piperidine-2-carboxamide

  • MDL number MFCD07322499
  • Cas Number : 1008692-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177...179
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71962

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71962
Availability details for
Quantity
  • Availability
  • Add to Cart