ENA311761092 Enamine

1-benzyl-N-(prop-2-en-1-yl)pyrrolidin-3-amine

  • MDL number MFCD11183217
  • Cas Number : 1039980-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71969

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71969
Availability details for
Quantity
  • Availability
  • Add to Cart