ENA311761100 Enamine

4-(3,4-dimethylphenyl)-2-fluorobenzoic acid

  • MDL number MFCD12859487
  • Cas Number : 1183614-10-9
  • Molecular Weight : 244.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-71990

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71990
Availability details for
Quantity
  • Availability
  • Add to Cart