ENA311761104 Enamine

2-amino-1-[3-(trifluoromethyl)phenyl]ethan-1-ol hydrochloride

  • MDL number MFCD01664408
  • Cas Number : 62064-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71994

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71994
Availability details for
Quantity
  • Availability
  • Add to Cart