ENA311761107 Enamine

3-(4-chlorobenzoyl)pyridine

  • MDL number MFCD00065043
  • Cas Number : 14548-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71997

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71997
Availability details for
Quantity
  • Availability
  • Add to Cart