ENA311761112 Enamine

2-methanesulfonylethane-1-thiol

  • MDL number MFCD12780441
  • Cas Number : 24517-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72003

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72003
Availability details for
Quantity
  • Availability
  • Add to Cart