ENA311761115 Enamine

[5-fluoro-2-(2-methyl-1H-imidazol-1-yl)phenyl]methanamine

  • MDL number MFCD11640259
  • Cas Number : 1096895-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72006

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72006
Availability details for
Quantity
  • Availability
  • Add to Cart