ENA311761117 Enamine

5-[(4-nitrophenyl)methyl]-1H-1,2,3,4-tetrazole

  • MDL number MFCD01240038
  • Cas Number : 131090-44-3
  • Molecular Weight : 205.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-71607

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71607
Availability details for
Quantity
  • Availability
  • Add to Cart