ENA311761119 Enamine

3-(2-hydroxyethyl)-2,3-dihydro-1,3-benzoxazol-2-one

  • MDL number MFCD00995481
  • Cas Number : 26726-75-0
  • Molecular Weight : 179.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72010

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72010
Availability details for
Quantity
  • Availability
  • Add to Cart