ENA311761131 Enamine

3-[(methoxycarbonyl)amino]propanoic acid

  • MDL number MFCD09962325
  • Cas Number : 158213-81-1
  • Molecular Weight : 147.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-71558

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71558
Availability details for
Quantity
  • Availability
  • Add to Cart