ENA311761136 Enamine

3-acetyl-4-hydroxybenzoic acid

  • MDL number MFCD00532680
  • Cas Number : 16357-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72031

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72031
Availability details for
Quantity
  • Availability
  • Add to Cart