ENA311761144 Enamine

3-tert-butyl-1,2,4-oxadiazol-5-ol

  • MDL number MFCD18089446
  • Cas Number : 163619-95-2
  • Molecular Weight : 142.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72039

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72039
Availability details for
Quantity
  • Availability
  • Add to Cart