ENA311761147 Enamine

2-amino-3-methylbutanamide hydrochloride

  • MDL number MFCD13562354
  • Cas Number : 93169-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 244...246
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72043

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72043
Availability details for
Quantity
  • Availability
  • Add to Cart