ENA311761148 Enamine

2-(4-hydroxypiperidin-1-yl)-N,N-dimethylacetamide

  • MDL number MFCD12145832
  • Cas Number : 1152845-38-9
  • Molecular Weight : 186.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72044

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72044
Availability details for
Quantity
  • Availability
  • Add to Cart