ENA311761149 Enamine

methyl 3-acetyl-4-hydroxybenzoate

  • MDL number MFCD06797942
  • Cas Number : 57009-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71618

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71618
Availability details for
Quantity
  • Availability
  • Add to Cart