ENA311761154 Enamine

pyrazolo[1,5-a]pyridine-3-carboxylic acid

  • MDL number MFCD00229214
  • Cas Number : 16205-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 223-224
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72058

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72058
Availability details for
Quantity
  • Availability
  • Add to Cart