ENA311761178 Enamine

1-(3,4-dichlorophenyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD11650174
  • Cas Number : 1152949-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71627
Availability details for
Quantity
  • Availability
  • Add to Cart