ENA311761183 Enamine

2-[2-methyl-5-(propan-2-yl)phenoxy]-2-phenylacetic acid

  • MDL number MFCD13719335
  • Cas Number : 1154658-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72089

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72089
Availability details for
Quantity
  • Availability
  • Add to Cart