ENA311761185 Enamine

3-[(1S)-1-hydroxyethyl]phenol

  • MDL number MFCD14705954
  • Cas Number : 194545-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71630
Availability details for
Quantity
  • Availability
  • Add to Cart