ENA311761194 Enamine

3-[(4-carbamoylpiperidin-1-yl)carbonyl]bicyclo[2.2.1]hept-5-ene-2-carboxylic acid

  • MDL number MFCD01812501
  • Cas Number : 1022736-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72096

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72096
Availability details for
Quantity
  • Availability
  • Add to Cart