ENA311761203 Enamine

1-{4-hydroxy-3-[(pyrrolidin-1-yl)methyl]phenyl}ethan-1-one

  • MDL number MFCD08445080
  • Cas Number : 92040-34-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-72104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72104
Availability details for
Quantity
  • Availability
  • Add to Cart