ENA313838946 Enamine

2-chloro-N-[2-(1-phenyl-1H-pyrazol-4-yl)ethyl]acetamide

  • MDL number MFCD18380581
  • Cas Number : 1269151-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-28337

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28337
Availability details for
Quantity
  • Availability
  • Add to Cart