ENA313838947 Enamine

N-{3-[1-(2-chloro-N-methylacetamido)ethyl]phenyl}benzamide

  • MDL number MFCD18380582
  • Cas Number : 1269151-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-28519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28519
Availability details for
Quantity
  • Availability
  • Add to Cart