ENA313838948 Enamine

2-chloro-N-[2,2-dimethyl-1-(thiophen-2-yl)propyl]acetamide

  • MDL number MFCD18380585
  • Cas Number : 1269151-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-47805

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47805
Availability details for
Quantity
  • Availability
  • Add to Cart