ENA313838950 Enamine

2-chloro-N-{[3-(2-oxopyrrolidin-1-yl)phenyl]methyl}acetamide

  • MDL number MFCD18380587
  • Cas Number : 1269152-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48002

Safety Information

msds https://www.enaminestore.com/catalog/EN300-48002
Availability details for
Quantity
  • Availability
  • Add to Cart