ENA313838951 Enamine

N-[(4-bromothiophen-2-yl)methyl]-2-chloroacetamide

  • MDL number MFCD18380588
  • Cas Number : 1269151-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-48190

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-48190
Availability details for
Quantity
  • Availability
  • Add to Cart