ENA313838954 Enamine

2-(2-chloroacetamido)-N-methyl-2-phenylacetamide

  • MDL number MFCD18380591
  • Cas Number : 502989-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-48860
Availability details for
Quantity
  • Availability
  • Add to Cart