ENA313838955 Enamine

3-(2-bromophenyl)butanoic acid

  • MDL number MFCD18380592
  • Cas Number : 66192-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61-62
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51827
Availability details for
Quantity
  • Availability
  • Add to Cart