ENA313838956 Enamine

2,2,2-trifluoroethyl N-[4-(4-methoxyphenyl)-1,3-thiazol-2-yl]carbamate

  • MDL number MFCD18380593
  • Cas Number : 1269151-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56604

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56604
Availability details for
Quantity
  • Availability
  • Add to Cart