ENA313838960 Enamine

3-(aminomethyl)piperidin-2-one hydrochloride

  • MDL number MFCD18380597
  • Cas Number : 1269151-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59757

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59757
Availability details for
Quantity
  • Availability
  • Add to Cart