ENA313838965 Enamine

2-(3-methylphenyl)-1,3-thiazole-4-sulfonyl chloride

  • MDL number MFCD18380604
  • Cas Number : 1269152-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71517

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71517
Availability details for
Quantity
  • Availability
  • Add to Cart