ENA313838971 Enamine

1-(2,2-dimethoxyethyl)-2-fluorobenzene

  • MDL number MFCD18380610
  • Cas Number : 1269151-82-7
  • Molecular Weight : 184.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72203

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72203
Availability details for
Quantity
  • Availability
  • Add to Cart