ENA313838973 Enamine

5-(3-methyl-1,2-oxazol-5-yl)-1,3-oxazole-4-carboxylic acid hydrochloride

  • MDL number MFCD18380612
  • Cas Number : 1269152-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72365

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72365
Availability details for
Quantity
  • Availability
  • Add to Cart